Learn More
Dräger™ Short-Term Detector Tubes: Diethyl Ether
Detector tubes
Brand: Dräger™ 6730501
Additional Details : Weight : 0.03000kg
Description
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Diethyl Ether | |
100 to 4000 | |
10 | |
59°F to 104°F | |
Diethyl ether 100/a |
3 to 15mg/L | |
10 | |
3 min. | |
15° to 40°C |