Learn More
Dräger™ Short-Term Detector Tubes: Benzene
Detector tubes
1040.00 NOK - 2325.00 NOK
Specifications
Components Measured Benzene
View More Specs
Products 5
Product Code Brand Components Measured Tube Type Measurement Range Humidity Range Price Quantity & Availability  
Product Code Brand Components Measured Tube Type Measurement Range Humidity Range Price Quantity & Availability  
10443291
View Documents Promotion Details
Dräger™
8101741
Benzene Benzene 15/a 15 to 420 1 to 30mg/L
1975.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 03-06-2024
to see stock.
10430102
View Documents Promotion Details
Dräger™
8101231
Benzene Benzene 2/a 2 to 60 1 to 15mg/L
2200.00 NOK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 03-06-2024
to see stock.
12304539
View Documents Promotion Details
Dräger™
6728071
Benzene Benzene 5/b 5 to 50 3 to 15mg/L
1915.00 NOK
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 03-06-2024
to see stock.
10743263
View Documents Promotion Details
Dräger™
8103691
Benzene Benzene 0.5/c 0.5 to 10 1 to 30mg/L
2325.00 NOK
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 03-06-2024
to see stock.
10553802
View Documents Promotion Details
Dräger™
6728561
Benzene Benzene 0.5/a 0.5 to 10 3 to 15mg/L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten. Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Specifications
Benzene