Learn More
Dräger™ Short-Term Detector Tubes: Sulfur Dioxide
Product Code. p-6467008
Change view
Click to view available options
Measurement Range:
0.1 to 3
0.5 to 25
1 to 25
20 to 2000
50 to 8000
Tube Type:
Sulfur dioxide 0.1/a
Sulfur dioxide 0.5/a2
Sulfur dioxide 1/a
Sulfur dioxide 20/a
Sulfur dioxide 50/b
Response Time:
20 min.
3 min.
3 to 6 min.
Not Applicable
Unit Size:
Pack of 10
Product Code. Measurement Range Tube Type Response Time unitSize
10482452 50 to 8000 Sulfur dioxide 50/b Not Applicable Pack of 10
10153312 20 to 2000 Sulfur dioxide 20/a 3 min. Pack of 10
10127390 0.5 to 25 Sulfur dioxide 0.5/a2 3 to 6 min. Pack of 10
10206731 1 to 25 Sulfur dioxide 1/a 3 min. Pack of 10
10632693 0.1 to 3 Sulfur dioxide 0.1/a 20 min. Pack of 10
5 options
Product Code. 10482452
Brand: Dräger™ 8101531
Detector tubes
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note:
Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Components Measured Sulfur Dioxide
Humidity Range Not Applicable
Measurement Range 50 to 8000
No. of Strokes 1/10
Quantity 10
Response Time Not Applicable
Temperature Range (English) 32°F to 122°F
Temperature Range (Metric) 0°C to 50°C
Tube Type Sulfur dioxide 50/b
compliance-icons
Product Identifier
  • X10 Tube Sulfur Dioxide 50/b
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Supplemental information
  • MIXTURE LIST-Contains : sulphur dioxide