Learn More
Dräger™ SCBA Safety Mask
Product Code. 15169473
Change view
Click to view available options
For Use With (Equipment):
Compressed air breathing apparatus
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15169473 Compressed air breathing apparatus Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15169473 Supplier Dräger™ Supplier No. R53070
  • ONLINE EXCLUSIVE PRICE
    7015.00 NOK
    6095.29 NOK / Pack of 1
    Save 919.71 NOK
    13% Off
Estimated Shipment: 29-01-2027
Inner mask and harness made of high quality silicone
  • Panorama Nova Silicone P PC facepiece
  • Without neck strap
Certifications
CE marked
Specifications
Certifications/Compliance EN 136
Connection Type Push-in
Product Type Facepiece
For Use With (Application) Supplies quickly and safely vital breathing air if the oxygen content drops below 17% by volume
For Use With (Equipment) Compressed air breathing apparatus
Harness Style Silicone
Material Silicone
Size Medium