Learn More
Dräger™ CMS Chips: Trichloroethylene
Product Code. 10056001
Change view
Click to view available options
Measurement Range:
5 to 100
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range unitSize
10056001 5 to 100 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10056001 Supplier Dräger™ Supplier No. 6406320
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Trichloroethylen CMS Chip
Humidity Range 1 to 30mg/L
Measurement Range 5 to 100
Response Time 40 to 330 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C