Learn More
Dräger™ Accessories for Portable Single Gas Detection Instruments
Product Code. 13498299
Change view
Click to view available options
Height (English):
2 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (English) Unit Size
13498299 2 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13498299 Supplier Dräger™ Supplier No. 8103525
  • ONLINE EXCLUSIVE PRICE
    995.78 NOK
    878.06 NOK / Pack of 1
    Save 117.72 NOK
    11% Off
 Restricted Item
Estimated Shipment: 14-01-2027
For use with single gas detection
Specifications
Type Replacement Sulfur Trioxide Filter
Height (English) 2 in.
Height (Metric) 5.08 cm