missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPP8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12752-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the peptidase S9B family, a small family of dipeptidyl peptidases that are able to cleave peptide substrates at a prolyl bond. The encoded protein shares similarity with dipeptidyl peptidase IV in that it is ubiquitously expressed, and hydrolyzes the same substrates. These similarities suggest that, like dipeptidyl peptidase IV, this protein may play a role in T-cell activation and immune function. Alternatively spliced transcript variants encoding different isoforms have been described.Specifications
DPP8 | |
Polyclonal | |
Unconjugated | |
DPP8 | |
dipeptidyl peptidase 8, Dipeptidyl peptidase VIII, DP8, DPP VIII, DPP8, DPRP 1, DPRP1, MSTP135, MSTP141, Prolyl dipeptidase DPP8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
315758, 54878, 74388 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6V1X1, Q80YA7 | |
Dpp8 | |
DPP8 Fusion Protein Ag3262 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |