Learn More
DPEP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16466-1-AP-150UL
This item is not returnable.
View return policy
Description
Probable metalloprotease which hydrolyzes leukotriene D4 (LTD4) into leukotriene E4 (LTE4).Specifications
| DPEP2 | |
| Polyclonal | |
| Unconjugated | |
| DPEP2 | |
| dipeptidase 2, DPEP2, MBD2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 291984, 319446, 64174 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5M872, Q8C255, Q9H4A9 | |
| Dpep2 | |
| DPEP2 Fusion Protein Ag9545 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |