missing translation for 'onlineSavingsMsg'
Learn More

DPEP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17167583
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17167583 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17167583 Supplier Invitrogen Supplier No. PA5109654

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

DPEP2 Polyclonal Antibody

DPEP2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen DPEP2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.96 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DPEP2
Gene Accession No. Q5M872, Q8C255, Q9H4A9
Gene Alias dipeptidase 2; DPEP2; F630103D06Rik; MBD2; MBD-2; Membrane-bound dipeptidase 2; putative membrane-bound dipeptidase 2; UNQ284/PRO323
Gene Symbols DPEP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 400-463 of human DPEP2 (NP_071750.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291984, 319446, 64174
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.