Learn More
DPD 1 blue buffer solution, Lovibond™
DPD 1 blue buffer solution
Brand:  Lovibond Tintometer 471016
Additional Details : Weight : 0.10000kg
Product Code. 10756723
  • 745.00 NOK / Pack of 6
Estimated Shipment: 30-07-2024
Description
DPD 1 blue buffer solution
DPD 1 blue buffer solution 6x15mL dropper bottle for approximately 300 tests 0.02-4mg/L chlorine. 
Specifications
Specifications
Buffer Solution