missing translation for 'onlineSavingsMsg'
Learn More

Melford DPBS 1X Solution

Product Code. 30018231
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018231 500 mL 500mL
30018230 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018231 Supplier Melford Supplier No. D22150500.0
 Restricted Item
Estimated Shipment: 13-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Phosphate Buffered Saline is a buffer solution commonly used in biological research.

Phosphate Buffered Saline is a buffer solution commonly used in biological research. PBS has many uses because it is isotonic and non-toxic to most cells. These uses include substance dilution and cell container rinsing.

Ready to use sterile 1X solution.

Ingredients:

Sodium Chloride: 137 mM

Potassium Chloride: 2.7 mM

Phosphate Buffer: 11.9 mM

pH: 7.4

Does not contain Calcium or Magnesium

Specifications

Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.