Learn More
Doxycycline (hyclate), MedChemExpress
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
Doxycycline hyclate, an antibiotic, is an orally active and broad-spectrum metalloproteinase (MMP) inhibitor. Doxycycline hyclate shows antibacterial activity and anti-cancer cell proliferation activity.
Specifications
Specifications
| Chemical Name or Material | Doxycycline (hyclate) |
| CAS | 24390-14-5 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C22H24N2O8.1/2C2H6O.ClH.1/2H2O |
| Quantity | 5 g |
| Synonym | Doxycycline hydrochloride hemiethanolate hemihydrate WC2031 |
| Solubility Information | DMSO : 240 mg/mL (467.89 mM; Need ultrasonic) ∣H2O : 125 mg/mL (243.69 mM; Need ultrasonic) |
| SMILES | O=C(C(C1=O)=C(O)[C@@H](N(C)C)[C@]2([H])[C@@H](O)[C@]3([H])[C@@H](C)C4=C(C(C3=C(O)[C@@]21O)=O)C(O)=CC=C4)N.Cl.[0.5H2O].[0.5C2H6O] |
| Molecular Weight (g/mol) | 512.94 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.