Learn More
Dowex™ 1X2, 100-200 mesh, ion-exchange resin
Product Code. 10432781 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
Product Code. Quantity unitSize
10432781 100 g 100g
10646232 500 g 500g
10424935 2.5 kg 2.5kg
3 options
Product Code. 10432781
Brand: Thermo Scientific Acros 202961000
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 9085-42-1 | C29H34ClN | 432.05 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 9085-42-1
Molecular Formula C29H34ClN
Molecular Weight (g/mol) 432.05
MDL Number MFCD00132715
InChI Key BBQMUEOYPPPODD-UHFFFAOYSA-M
Synonym dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
PubChem CID 16212807
IUPAC Name 1,4-diethenylbenzene 4-ethenyl-N,N,N-trimethylanilinium ethenylbenzene chloride
SMILES [Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1
Specifications
Physical Form Beads
Particle Size Particle size analysis: on 50 mesh (cumulative): < 15%, through 140 mesh: < 15%
Packaging Plastic Bottle
Volume Total Capacity wet volume capacity, Cl form (meq/mL):> 0.6
Water Retention Capacity Cl form: 70 to 80%
Fieser 01,511; 04,267; 06,302; 07,182; 08,263
Merck Index 10, 2182
Quantity 100 g
Solubility Information Solubility in water: insoluble.
Chemical Name or Material Dowex™ 1X2