Learn More
SymLine™ Funnel Adapter
Product Code. 15254708
Change view
Click to view available options
Closure Size:
GL14, R 2 in. BSP/G 2 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size Unit Size
15254708 GL14, R 2 in. BSP/G 2 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15254708 Supplier SymLine™ Supplier No. 107885
  • ONLINE EXCLUSIVE PRICE
    3230.00 NOK
    2981.16 NOK / Pack of 1
    Save 248.84 NOK
    7% Off
Estimated Shipment: 29-01-2027
Dovetail your SymLine™ System to suit your exact needs. Here, you will find many useful ancillaries.
SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.
  • Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
  • All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards
Specifications
Closure Size GL14, R 2 in. BSP/G 2 in.
Closure Color Black
Description for Funnel with Mechanical Level Indicator
Color Black
Liner Material HDPE-EC
For Use With (Equipment) Funnel
Material HDPE-EC
Product Type Funnel Adapter