Learn More
SymLine™ Thread Adapter, Type 11
Product Code. 10364260
Change view
Click to view available options
Closure Size:
GL25, G 1/2 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size Unit Size
10364260 GL25, G 1/2 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10364260 Supplier SymLine™ Supplier No. 106415
  • ONLINE EXCLUSIVE PRICE
    496.00 NOK
    444.80 NOK / Pack of 1
    Save 51.20 NOK
    10% Off
View all promotions
 Restricted Item
Estimated Shipment: 27-11-2026
Dovetail your SymLine™ System to suit your exact needs. Here, you will find many useful ancillaries.
SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.
  • Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
  • All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards
Specifications
Closure Size GL25, G 1/2 in.
Closure Color Black
Design External Thread
Description GL25 (m) to G1/2 in. (m)
Color Black
Liner Material HDPE-EC
Material HDPE-EC
Product Type Thread Adapter