Learn More
Usbeck™ Double Spoon
Product Code. 10380842
Change view
Click to view available options
Length (Metric):
120 mm
180 mm
250 mm
300 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
10380842 120 mm Each
10025491 180 mm Each
10127201 250 mm Each
10137201 300 mm Each
4 options
Product Code. 10380842
Brand: Usbeck™ 3351
Only null left
Add to basket
Add to basket
Measure and transfer chemical with precision. Usbeck™ Double Spoon is ideal for measuring and transferring chemicals.
  • Stainless steel
  • Rigid
  • Round handle
Specifications
Product Type Double Spoon End Spatula
Width (Metric) Blade 17/25 mm
Material Stainless Steel
Length (Metric) 120 mm