Learn More
Cole-Parmer™ Double Open End Filter Cartridges, polypropylene
Use these filter cartridges for a chemically inert media with high dirt-loading capacity. Cole Parmer™ Polypropylene Double Open End Filter Cartridges are ideal for air/gas filtration.
Brand:  Cole-Parmer™ 06479-32
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11796109
  • 1935.00 NOK / Each
Estimated Shipment: 15-07-2024
Description
  • Autoclavable to 121°C and in-line sterilizable to 125°C for up to 30-minute cycles
  • Filters can be sanitized with compatible chemical agents
  • Cartridges are suitable to 82°C at 10 psid
  • Cartridges comply with U.S. CFR Title 21 guidelines for repeated food contact, comply with USP Class VI, 121°C Plastics guidelines, and pass the MEM Elution Cytotoxicity Test
  • Aqueous extracts contain less than 0.25 EU/mL
  • Polypropylene Cartridges made of thermally bonded high-purity polypropylene fiber
  • Tolerant to an accumulated exposure of 10 hours
  • FDA-acceptable
  • Hydrophobic; to use with aqueous solutions, pre-wet with methanol, then rinse with water before use for liquid filtration
Specifications
Specifications
Autoclavable
1 μm
Filter Cartridge
Polypropylene
1 Pack
254 mm