Learn More
Hughes Double Nozzle Handheld Eye/Face/Body Wash
Product Code. 17782338
Change view
Click to view available options
Color:
Red
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17782338 Red Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17782338 Supplier Hughes Supplier No. HEWS5022RB
  • ONLINE EXCLUSIVE PRICE
    6938.74 NOK
    4019.33 NOK / Pack of 1
    Save 2919.41 NOK
    42% Off
Estimated Shipment: 17-12-2026
A wall mounted, handheld eye, face and body wash with two nozzles. Developed to suit hazardous environments where there is a risk of small spillages of chemicals or dangerous substances.
  • A wall mounted, handheld eye, face and body wash with two nozzles
  • Developed to suit hazardous environments where there is a risk of small spillages of chemicals or dangerous substances
  • 5 m of reinforced hose makes these handheld units particularly useful for treating disabled personnel
  • The lock-on actuating button for ease of use meets European and international standards
  • Fitted with dust covers, a non-return valve and a flow regulator
Specifications
Color Red
Height (Metric) 20 cm
Product Type Eye, Face and Body Combination Wash Stations
Width (Metric) 18 cm