Description
- Covers the full head, as required by the HACCP process
- Extra comfort with double elastic to ensure excellent retention
- Ability to use color code for tracking
- 12 g/m2 fabric weight
Specifications
Specifications
| Material | Polypropylene |
| Color | Green |
| Disposable | Single-use |
| Design Type | Clip Cap |
| Sterility | Non-sterile |
| Certifications/Compliance | EN ISO 9001 |
| Quantity | 250 Pk. |
| Dimensions (L x W) | 40 x 55 cm |
| Includes | Caps and Wall Dispenser |
| For Use With (Application) | Industrial, Laboratory |