Få mere at vide
Få mere at vide
Metro™ Double-Deep qwikTRAK™ Mobile Unit Kits
Klik for at se tilgængelige muligheder
Width (English):
18 in.
21 in.
24 in.
Width (English) Shelf:
18 in.
21 in.
24 in.
Finish:
Chrome
Heavy Polymer
Metroseal Green Epoxy
Stainless Steel
Pakningsstørrelse:
Each
Beskrivelse
- For use with Metro Super Erecta™, Super Erecta Pro™ Super Adjustable™ and MetroMax i™ shelving systems
- Kit includes eight 74 in. (188cm) high -UP posts, four V-groove casters; one stainless steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware
Ordreinfo
Order one kit per double-deep system. Shelves are sold separately.
Tekniske data
Tekniske data
| Product Type | Mobile Unit Kit |
| Finish | Heavy Polymer |
| For Use With (Equipment) | Metro™ MetroMax i™ shelving system |
| Includes | Eight 74 in. high up posts, four V-groove casters, one Stainless Steel center channel caster assembly, donut bumpers, tie bar brackets and other hardware |
| Width (English) Shelf | 18 in. |
| Width (English) | 18 in. |
| Width (Metric) Shelf | 45.7 cm |
| Width (Metric) | 45.7 cm |
| Material | Polymer |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion