missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Double Bagged - USP Class VI PharmaScoops™
BIOPHARMA

Product Code. 17554696 Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
125 mL
250 mL
1000 mL
Unit Size:
Pack of 10
Pack of 25
Pack of 40
Pack of 50
This item is not returnable. View return policy

Product Code. 17554696

Brand: Sampling Systems™ 9045H250S

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Double Bagged PharmaScoops™ are made from high grade medical USP Class VI HDPE. They are available in a range of sizes from 125 mL to 1000 mL. This premium range is double bagged. A box liner is also used giving a third layer of protection.

  • These sterile scoops are commonly used in the manufacture and sampling of cosmetics, food and pharmaceuticals.
  • These scoops all have an ergonomic handle which is positioned high up the body so as to keep the hand as far away from the powder as possible.
  • The scoops are designed to be used straight from the bag.
  • The double bagging means that the outer bag can be removed whilst entering the cleanroom, ensuring the cleanliness inside the cleanroom or manufacturing area.
  • Made from food grade, FDA and EC compliant HDPE.
  • HDPE has good chemical resistance and can be easily recycled.
  • The funnels are sterilised by gamma irradiation.
  • USP Class VI - High quality medical grade polymer that has also been independently tested to USP 661.
  • The scoops are entirely made in cleanroom conditions with final packing carried out in a class 10,000 cleanroom.
  • Each scoop is 100% inspected before packing.

Specifications

Sterility Sterile
Capacity (Metric) 250 mL
Material HDPE
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.