Learn More
Kartell™ Dosipet B
Product Code. 10172761
Click to view available options
Volume (Metric):
2.5 to 25 mL
5 to 50 mL
Increments:
0.5 mL
1 mL
Unit Size:
Each
Product Code. 10172761
Brand: Kartell™ 8305000
Only null left
Add to basket
Add to basket
Accurately and efficiently dispense solutions. Kartell™ Dosipet B can be used with almost every water solution type and comes equipped with a cylinder and piston.
  • Can be used with every water solution type
  • Do not use with solution containing hydrofluoric acid
  • Marked by a high degree of convenience and reliability
  • Equipped with a 3.3 borosilicate glass cylinder and PTFE piston
Specifications
Product Type Dosipet 50
Certifications/Compliance CE marked / IVD certified
Description GL45 Thread Size
Volume (Metric) 5 to 50 mL
For Use With (Application) Water solution type
Includes 3.3 Borosilicate Glass cylinder and PTFE piston
Increments 1 mL
Model Dosipet 50
Quantity 1 Pack
Thread Size GL 45