Learn More
Lenz Laborglasintrumente Dosing Funnel
Product Code. 11993601
Change view
Click to view available options
Capacity (Metric):
1 L
4 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11993601 1 L Each
11903611 4 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11993601 Supplier Lenz Laborglasintrumente Supplier No. 06350070
  • ONLINE EXCLUSIVE PRICE
    2110.00 NOK
    1895.52 NOK / Pack of 1
    Save 214.48 NOK
    10% Off
Estimated Shipment: 26-01-2027
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • System contents and spare parts for one-way reaction/distillation units 6, 10, 20 l, LF 150
Specifications
Capacity (Metric) 1 L
For Use With (Equipment) 6 and 10 L reaction vessel/distillation unit
Joint Number NS 29/32