Learn More
Dorzolamide Hydrochloride, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Dorzolamide Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Dorzolamide Hydrochloride |
| CAS | 130693-82-2 |
| Analyte or Component Names | Dorzolamide Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H16 N2 O4 S3 . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Dorzolamide Hydrochloride |
| InChI Formula | InChI=1S/C10H16N2O4S3.ClH/c1-3-12-8-4-6(2)18(13,14)10-7(8)5-9(17-10)19(11,15)16;/h5-6,8,12H,3-4H2,1-2H3,(H2,11,15,16);1H/t6-,8-;/m0./s1 |
| SMILES | Cl.CCN[C@H]1C[C@H](C)S(=O)(=O)c2sc(cc12)S(=O)(=O)N |
| IUPAC Name | (4S,6S)-4-(ethylamino)-6-methyl-7,7-dioxo-5,6-dihydro-4H-thieno[2,3-b]thiopyran-2-sulfonamide;hydrochloride |
| Show More |