Learn More
Thermo Scientific™ Stainless Steel Door Glass Covers
Product Code. 15593751 Shop All Thermo Scientific Products
Change view
Click to view available options
Installation:
Customer Installed
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Installation Unit Size
15593751 Customer Installed Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15593751 Supplier Thermo Scientific™ Supplier No. 190892
  • ONLINE EXCLUSIVE PRICE
    7510.00 NOK
    7019.27 NOK / Pack of 1
    Save 490.73 NOK
    6% Off
 Restricted Item
Estimated Shipment: 28-01-2027
For Use With: Environmental Chamber (3948 and 3962)
Customize environmental chamber and incubator configurations to better suit the specific needs of the lab with Thermo Scientific™ Door Options.
TRUSTED_SUSTAINABILITY
Specifications
Description Stainless Steel Door Glass Covers
For Use With (Equipment) Environmental Chamber (3948 and 3962)
Lighting LED
Material Stainless Steel
Type Stainless Steel Door Glass Covers
Installation Customer Installed