Learn More
BINDER™ Door Lock for Series KBF-S, KBF-S ECO
Product Code. 17333673 Shop All BINDER Products
Change view
Click to view available options
Material:
Plastic
Unit Size:
Each
Product Code. Material unitSize
17333673 Plastic Each
1 options
Product Code. 17333673
Brand: BINDER™ 80122087
Only null left
Add to basket
Add to basket
BINDER™ lockable door handle is an option for use with Constant Climate Chambers KBF-S and KBF-S ECO
  • Door lock
  • Lockable door handle
Specifications
Description Door lock - lockable door handle
For Use With (Equipment) BINDER Chambers KBF-S 720, KBF-S 1020, KBF-S ECO 720, KBF-S ECO 1020
Material Plastic
Type Door Lock