Learn More
Donkey anti-Rat IgG (H+L) Secondary Antibody, DyLight 405, Novus Biologicals™
Product Code. 18508210 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. Quantity unitSize
18508210 1 mg 1mg
1 options
Product Code. 18508210
Brand: Novus Biologicals NBP175651
Donkey Polyclonal Antibody
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rat samples. It is validated for Flow Cytometry, Immunomicroscopy
TRUSTED_SUSTAINABILITY
Specifications
Antigen IgG (H+L)
Applications Flow Cytometry, Immunomicroscopy
Classification Polyclonal
Concentration LYOPH
Conjugate DyLight 405
Dilution Flow Cytometry 1:20 - 1:2000, Immunomicroscopy 1:20 - 1:2000
Formulation 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
Host Species Donkey
Immunogen This Donkey anti-Rat IgG (H+L) Secondary Antibody [DyLight 405] was developed against purified rat IgG (H&L).
Purification Method Affinity purified
Quantity 1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Reconstitution Reconstitute with sterilized deionized water. Centrifuge to remove any particulates.
Target Species Rat
Content And Storage Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage.
Form Purified
Isotype IgG