Learn More
Thermo Scientific™ Domed PCR Caps, strips of 12
Descrizione
Ensure proper sealing on all PCR strip tubes and 96-well PCR plates with precision molded Thermo Scientific™ PCR Plate Cap Strips.
Domed PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Highlights:
- Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
- Large end tabs for strip removal (12 strips only)
Certifications:
Certified free from human DNA, RNases, and DNases.
Specifica
Specifica
| Color | Clear |
| Material | Polypropylene |
| No. of Wells | 12 |
| Quantity | 10 packs of 8 |
| Format | Strips of 12 Domed Caps |
| Product Type | PCR Plate Cap Strip |
| Certified RNase/DNase-free | Yes |
| Plate Compatibility | 96-well Plate |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.