Learn More
TFA Dostmann™ Domatic Baro Thermo Hygrometer
Product Code. 11852623
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
11852623 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11852623 Supplier TFA Dostmann™ Supplier No. 20.3006.42
  • ONLINE EXCLUSIVE PRICE
    440.91 NOK
    325.60 NOK / Pack of 1
    Save 115.31 NOK
    26% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Specifications
Includes Barometer, thermometer and hygrometer
Height (Metric) 147 mm
Material Plastic
Weight (Metric) 200 g
Diameter (Metric) Face 136 mm