Learn More
Dolutegravir Sodium, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Dolutegravir Sodium
Specifications
Specifications
| Chemical Name or Material | Dolutegravir Sodium Salt |
| CAS | 1051375-19-9 |
| Analyte or Component Names | Dolutegravir Sodium Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H18 F2 N3 O5 . Na |
| Certifications/Compliance | ISO 17034 |
| Synonym | 2H-Pyrido[1',2':4,5]pyrazino[2,1-b][1,3]oxazine-9-carboxamide, N-[(2,4-difluorophenyl)methyl]-3,4,6,8,12,12a-hexahydro-7-hydroxy-4-methyl-6,8-dioxo-, sodium salt (1:1), (4R,12aS)-, Dolutegravir sodium, Dolutegravir sodium salt, GSK 1349572A, GSK 1349572A |
| InChI Formula | InChI=1S/C20H19F2N3O5.Na/c1-10-4-5-30-15-9-24-8-13(17(26)18(27)16(24)20(29)25(10)15)19(28)23-7-11-2-3-12(21)6-14(11)22;/h2-3,6,8,10,15,27H,4-5,7,9H2,1H3,(H,23,28);/q;+1/p-1/t10-,15+;/m1./s1 |
| SMILES | [Na+].C[C@@H]1CCO[C@H]2CN3C=C(C(=O)NCc4ccc(F)cc4F)C(=O)C(=C3C(=O)N12)[O-] |
| IUPAC Name | sodium;(4R,12aS)-9-[(2,4-difluorophenyl)methylcarbamoyl]-4-methyl-6,8-dioxo-3,4,12,12a-tetrahydro-2H-pyrido[5,6]pyrazino[2,6-b][1,3]oxazin-7-olate |
| Show More |