missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The protein encoded by this gene is a member of the DOK family of membrane proteins, which are adapter proteins involved in signal transduction. The encoded protein interacts with phosphorylated receptor tyrosine kinases to mediate neurite outgrowth and activation of the MAP kinase pathway. In contrast to other DOK family proteins, this protein does not interact with RASGAP.
Specifications
Specifications
| Antigen | DOK5 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.27 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | DOK5 |
| Gene Accession No. | Q91ZM9, Q9P104 |
| Gene Alias | C20orf180, docking protein 5, DOK5, Insulin receptor substrate 6, IRS 6, IRS6 |
| Gene Symbols | DOK5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?