missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOK5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11948-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the DOK family of membrane proteins, which are adapter proteins involved in signal transduction. The encoded protein interacts with phosphorylated receptor tyrosine kinases to mediate neurite outgrowth and activation of the MAP kinase pathway. In contrast to other DOK family proteins, this protein does not interact with RASGAP.Specifications
| DOK5 | |
| Polyclonal | |
| Unconjugated | |
| DOK5 | |
| C20orf180, docking protein 5, DOK5, Insulin receptor substrate 6, IRS 6, IRS6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 502694, 55816, 76829 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q91ZM9, Q9P104 | |
| DOK5 | |
| DOK5 Fusion Protein Ag2615 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction