missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOC2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20574-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to DOC2B.
There are at least two protein isoforms of the Double C2 protein, namely alpha and beta , which contain two C2-like domains. DOC2A and DOC2B are encoded by different genes; these genes are at times confused with the unrelated DAB2 gene which was initially named DOC-2. DOC2B is expressed ubiquitously and is suggested to be involved in Ca-dependent intracellular vesicle trafficking in various types of cells.Specifications
| DOC2B | |
| Polyclonal | |
| Unconjugated | |
| DOC2B | |
| Doc2 beta, DOC2B, DOC2BL, double C2 like domains, beta | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13447, 81820, 8447 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P70169, P70610, Q14184 | |
| Doc2b | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction