missing translation for 'onlineSavingsMsg'
Learn More

DO3A tert-Butyl ester, MedChemExpress

Product Code. p-200099223
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
Unit Size:
100mg
200mg
This item is not returnable. View return policy

Product Code. 16495946

Brand: Medchem Express HY12692

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress DOTA tert-Butyl ester is a benxyl derivative of the cyclic tosamide; can be nitrated directly; is more convenient to incorporate the nitro group after deprotection lithium aluminum hydride.

DOTA tert-Butyl ester is a benxyl derivative of the cyclic tosamide

Specifications

Chemical Name or Material DO3A tert-Butyl ester
Physical Form Solid
CAS 122555-91-3
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C26H50N4O6
Synonym DO3A tert-butyl DO3A-t-Bu-ester
Solubility Information DMSO : 62.5 mg/mL (121.43 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(OC(C)(C)C)CN1CCN(CC(OC(C)(C)C)=O)CCNCCN(CC(OC(C)(C)C)=O)CC1
Molecular Weight (g/mol) 514.7
Formula Weight 514.7
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.