missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNMT3B Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | DNMT3B |
|---|---|
| Concentration | 467 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16805295
|
Proteintech
26971-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16895285
|
Proteintech
26971-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The DNMT3B antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human DNMT3B. This antibody recognizes human, mouse antigen. The DNMT3B antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| DNMT3B | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Human, Mouse | |
| IgG |
| 467 μg/mL | |
| Polyclonal | |
| Rabbit | |
| 1789 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title