missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DNMT2 (Human) Recombinant Protein
Human DNMT2 full-length ORF ( AAH47733, 1 a.a. - 391 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001787-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MEPLRVLELYSGVGGMHHALRESCIPAQVVAAIDVNTVANEVYKYNFHHTQLLAKTIEGITLEEFDRLSFDMILMSPPCQPFTRIGRQGDMTDSRTNSFLYILDILPRLQKLPKYILLENVKGFEVSSTRDLLIQTIENCGFQYQEFLLSPTSLGIPNSRLRYFLIAKLQSEPLPFQAPGQVLMEFPKIESVHPQKYAMDVEEKIQEKNVEPNISFDGSIQCSGKDAILFKLETAEEIHRKNQQDSDLSVKMLKDFLEDDTDVNQYLLPPKSLLRYALLLDIVQPTCRRSVCFTKGYGSYIEGTGSVLQTAEDVQVENIYKSLTNLSQEEQITKLLILKLRYFTPKEIANLLGFPPEFGFPEKITVKQRYRLLGNSLNVHVVAKLIKILYE
Specifications
AAH47733 | |
tRNA aspartic acid methyltransferase 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TRDMT1 | |
GST |
1787 | |
Wheat germ expression system | |
25 μg | |
DNMT2, M.HsaIIP, PuMet, RNMT1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |