missing translation for 'onlineSavingsMsg'
Learn More

DNCIC1 Antibody [Alexa Fluor« 594], Novus Biologicals Biologicals™

Product Code. 30499724 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity unitSize
30499724 0.1 mL 0.1mL
1 options
This item is not returnable. View return policy

Product Code. 30499724

Brand: Novus Biologicals NBP338053AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DNCIC1 Polyclonal antibody specifically detects DNCIC1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNCIC1
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias cytoplasmic dynein 1 intermediate chain 1, Cytoplasmic dynein intermediate chain 1, DNCI1cytoplasmic, intermediate polypeptide 1, DNCIC1DH IC-1, Dynein intermediate chain 1, cytosolic, dynein, cytoplasmic 1, intermediate chain 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human DNCIC1 (NP_001129028.1).,, Sequence:, MSDKSDLKAELERKKQRLAQIREEKKRKEEERKKKEADMQQKKEPVQDDSDLDRKRRETEALLQSIGISPEPPLVPTPMSPSSKSVSTPSEAGSQDSGDLGPLTRTLQWDTDPSVLQLQSDSELGRRLHKLGVSKVTQVDFLPREVVSYSKETQTPLATHQSEEDEEDEEMVESKVGQDSELENQDKKQEVKEAPPRELTEEEKQQILHSEEFLIFFDRT
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Golgi Apparatus Markers, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1780
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.