Learn More
Thermo Scientific™ DNAPac™ PA100 Oligonucleotide HPLC Columns
Product Code. 11381723
Click to view available options
Length (Metric):
50 mm
250 mm
Diameter (Metric):
2 mm
4 mm
9 mm
22 mm
Pakningsstørrelse:
Each
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 11381723
Brand: Thermo Scientific™ 043018
Kun null tilbage
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy
Features strong anion-exchange resin for high-resolution analysis and purification of synthetic and modified oligonucleotides.
Feature strong anion-exchange resin for high-resolution analysis and purification of synthetic and modified oligonucleotides with Thermo Scientific™ Dionex™ DNAPac™ PA100 Oligonucleotide Columns.
  • 13μm pellicular, nonporous polymeric resin with bound quaternary amine-functionalized MicroBeads™
  • Capable of n, n-1 resolution for oligonucleotides
  • Resolves oligonucleotides with secondary structures
  • Compatible with solvent, high pH, and high temperatures
  • Analyzes phosphorothioate-based clinical samples
  • Provides easy scale-up
TRUSTED_SUSTAINABILITY
Tekniske data
Column Format Guard Column
Diameter (Metric) 4 mm
For Use With ASRS, DX-500 BioLC Chromatography System
Length (Metric) 50 mm
Max. Pressure 4000 psi (275 bar)
Particle Size 13 μm
pH 2 to 12.5
Pore Size Nonporous
Quantity Each
Temperature 90°C
Stationary Phase Quaternary Ammonium
Product Line DNAPac
Packing Material Polymeric Resin
Column Type Ion Exchange
Phase Ion Exchange
Type Guard Column
Technique Bio-LC
Green Features Responsibly packaged
Vis mere Vis mindre
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.