missing translation for 'onlineSavingsMsg'
Learn More

DNALI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16897534 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16897534 20 μL 20µL
16887534 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16897534

Brand: Proteintech 176011AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene is the human homolog of the Chlamydomonas inner dynein arm gene, p28. The precise function of this gene is not known, however, it is a potential candidate for immotile cilia syndrome (ICS). Ultrastructural defects of the inner dynein arms are seen in patients with ICS. Immotile mutant strains of Chlamydomonas, a biflagellated algae, exhibit similar defects.

Specifications

Antigen DNALI1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DNALI1
Gene Accession No. O14645, Q4FZV3, Q8BVN8
Gene Alias DNALI1, hp28, P28
Gene Symbols Dnali1
Host Species Rabbit
Immunogen DNALI1 Fusion Protein Ag11652
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 298524, 75563, 7802
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.