Learn More
DNAJB9 Rabbit anti-Human, Mouse, Rat, Biotin, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech BIOTIN-13157-100UL
This item is not returnable.
View return policy
Description
This gene is a member of the J protein family. J proteins function in many cellular processes by regulating the ATPase activity of 70 kDa heat shock proteins. This gene is a member of the type 2 subgroup of DnaJ proteins. The encoded protein is localized to the endoplasmic reticulum. This protein is induced by endoplasmic reticulum stress and plays a role in protecting stressed cells from apoptosis.Specifications
| DNAJB9 | |
| Polyclonal | |
| Biotin | |
| DNAJB9 | |
| DNAJB9, ERdj4, Mdg 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 24908, 27362, 4189 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.3 | |
| P97554, Q9QYI6, Q9UBS3 | |
| Dnajb9 | |
| DNAJB9 Fusion Protein Ag3789 | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |