Learn More
DNA2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21599-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to DNA2.
DNA2 is a conserved helicase/nuclease involved in themaintece of mitochondrial and nuclear DNA stability.Specifications
DNA2 | |
Polyclonal | |
Unconjugated | |
DNA2 | |
DNA2, DNA2L, KIAA0083 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1763 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P51530 | |
DNA2 | |
DNA2 Fusion Protein Ag16206 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |