Learn More
MP Biomedicals™ DNA-OFF™
Product Code. 11477130 Shop All MP Biomedicals Products
Change view
Click to view available options
Volume (Metric):
500 mL
Unit Size:
500mL
Product Code. Volume (Metric) unitSize
11477130 500 mL 500mL
1 options
Product Code. 11477130
Brand: MP Biomedicals™ undefined
DNA-OFF™ is a ready to use non-alkaline cleaning solution that is highly active against DNA contamination. It is non-carcinogenic, non corosive and heat resistant solution used for eliminating DNA from surfaces, such as PCR workstations, countertops, apparatus and pipettors.
  • The presence of contaminant DNA at a PCR work area can result in DNA amplification artifacts.
  • This ready-to-use solution is highly active against DNA contamination and is intended for use at PCR workstations, including countertops, apparatus and pipettors.
  • It is stable, heat resistant, non-corrosive and free of carcinogenic compounds.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Removal Agent
Volume (English) 16.9 oz.
Volume (Metric) 500 mL
For Use With (Application) Highly active non-alkaline coleaning solution for DNA contamination, ready to use.
For Use With (Equipment) Intended for use at PCR workstations, including countertops, apparatus and pipettors
Packaging Type Bottle