Learn More
VACUUBRAND™ DN 16/DN 10mm Polypropylene Small Flange With Hose Nozzle
Click to view available options
Material:
Polypropylene
Unit Size:
Each
Specifications
Specifications
| Material | Polypropylene |
| Quantity | 1 Each |
| Size | DN 16/DN 10 mm |
| Type | Small Flange |