missing translation for 'onlineSavingsMsg'
Learn More

DMRTB1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26118-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16894645

  • 3468.15 NOK / 150µL
Estimated Shipment: 16-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

DMRTB1 gene ontology annotations related to this gene include DNA-binding transcription factor activity.
Specifications

Specifications

DMRTB1
Polyclonal
Unconjugated
DMRTB1
DMRTB1
Rabbit
Antigen Affinity Chromatography
RUO
63948
-20°C
Liquid
Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96MA1
DMRTB1
DMRTB1 Fusion Protein Ag23639
150 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers