missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMRT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14313-1-AP-20UL
This item is not returnable.
View return policy
Description
DMRT1 Polyclonal antibody apecifically detetects DMRT1 antigen in Human, Mouse, Rat Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.Specifications
| DMRT1 | |
| Polyclonal | |
| Unconjugated | |
| BC040847 | |
| Rabbit | |
| Antigen affinity purification | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 500 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| DMRT1 | |
| DMRT1 fusion protein Ag5547 | |
| 20 μL | |
| 1761 | |
| Store at -20°C. Stable for one year after shipment. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction