missing translation for 'onlineSavingsMsg'
Learn More

DMRT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14313-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 17699596

  • 3828.53 NOK / 150µL
Estimated Shipment: 28-09-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

DMRT1 Polyclonal antibody apecifically detetects DMRT1 antigen in Human, Mouse, Rat Samples. It is validated for Western Blot, Immunohistochemistry, ELISA.
Specifications

Specifications

DMRT1
Polyclonal
Unconjugated
BC040847
Rabbit
Antigen affinity purification
Primary
Human, Mouse, Rat
Liquid
Western Blot, Immunohistochemistry, ELISA
500 μg/mL
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
DMRT1
DMRT1 fusion protein Ag5547
150 μL
1761
Store at -20°C. Stable for one year after shipment.
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers