Learn More
Gibco™ DMEM/F-12, no phenol red
GREENER_CHOICE
Product Code. 12088089
Change view
Click to view available options
Quantity:
10 x 500 mL
Unit Size:
5000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12088089 10 x 500 mL 5000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12088089 Supplier Gibco™ Supplier No. 21041033
Edge
DMEM/F-12, no phenol red
DMEM/F-12 (Dulbecco's Modified Eagle Medium/Nutrient Mixture F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM/F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of DMEM/F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
Using DMEM/F-12
DMEM/F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids and vitamins with F-12's wide variety of components. DMEM/F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM/F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM/F-12 uses a sodium bicarbonate buffer system and therefore requires a 5–10% CO2 environment to maintain physiological pH.
cGMP manufacturing and quality system
For supply chain continuity, we manufacture DMEM/F-12 at two separate facilities, located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP-manufacturing requirements, are certified to ISO 13485, and are registered with the FDA as medical device manufacturers.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 1 X
Form Liquid
Product Type DMEM Medium
With Additives Glutamine, Sodium Pyruvate
Without Additives No HEPES, No Phenol Red
For Use With (Application) Mammalian cell culture
Quantity 10 x 500 mL
Certifications/Compliance ISO 13485 standards, FDA
pH 7
Content And Storage 2 to 8°C
Description DMEM Medium, 500mL, 1 X Concentration, Liquid Form, With L-glutamine, Sodium Pyruvate, 7 pH, 12 Months Shelf Life, 2 to 8°C Storage Conditions, Soluble in Water, Used for Mammalian Cell Culture