Learn More
Gibco™ DMEM
Product Code. 16219961
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16219961 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16219961 Supplier Gibco™ Supplier No. 670087
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Dulbecco's Modified Eagle's Medium (DMEM)
A high-glucose formulation and contains phenol red, 4 mM L-glutamine, 4.5 g/L glucose, 1 mM sodium pyruvate, and 1.5 g/L sodium bicarbonate.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Protect from light, 2°C to 8°C
Form Liquid
Product Type Basal Media
For Use With (Application) Cell Culture
Quantity 500 mL