missing translation for 'onlineSavingsMsg'
Learn More

DLX5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10592-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16847073

  • 3828.53 NOK / 150µL
Estimated Shipment: 10-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of a homeobox transcription factor gene family similar to the Drosophila distal-less gene. The encoded protein may play a role in bone development and fracture healing. Mutation in this gene, which is located in a tail-to-tail configuration with another member of the family on the long arm of chromosome 7, may be associated with split-hand/split-foot malformation.
Specifications

Specifications

DLX5
Polyclonal
Unconjugated
DLX5
distal less homeobox 5, DLX5, Homeobox protein DLX 5
Rabbit
Antigen Affinity Chromatography
RUO
13395, 1749, 25431
-20°C
Liquid
Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P50575, P56178, P70396
DLX5
DLX5 Fusion Protein Ag0910
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers