Learn More
DLK1 Mouse anti-Human, Unconjugated, Clone: 2C3H8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67923-1-IG-20UL
Additional Details : Weight : 0.00020kg
Description
The DLK1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human DLK1. This antibody recognizes human, pig antigen. The DLK1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
DLK1 | |
Monoclonal | |
Unconjugated | |
20 μL | |
8788 | |
IgG1 |
Western Blot, ELISA | |
2C3H8 | |
Mouse | |
Primary | |
Human, Pig |